missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Vps34-IN1

Vps34 inhibitor

Encompass_Preferred

554.85 DKK - 1960.20 DKK

Specifications

CAS 1383716-33-3
Molecular Formula C21H24ClN70
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17109981
Encompass_Preferred
View Documents
STEMCELL Technologies
100-0255
5 mg
1960.20 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17199971
Encompass_Preferred
View Documents
STEMCELL Technologies
100-0254
1 mg
554.85 DKK
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Vps34-IN1 is a potent and reversible inhibitor (IC50 = 25 nM) of vacuolar protein sorting 34 (Vps34), a class II phosphoinositide 3-kinase that regulates autophagosome formation (Bago et al.)
  • Reduces accumulation of phosphorylated phosphatidylinositol (PtdIns(3)P) at endosomal membranes (Bago et al.)
Specifications

Specifications

1383716-33-3
C21H24ClN70
≥98%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies Vps34-IN1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.