missing translation for 'onlineSavingsMsg'
Learn More
Vps34-IN-1, MedChemExpress Product Code.: 16498067

Vps34-IN-1, MedChemExpress

Product Code. p-200101946
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498067

Brand: Medchem Express HY12795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vps34-IN-1 is an inhibitor of Vps34 extracted from patent WO2012085815A1, compound example 16a, with an IC50 of 4 nM. Vps34-IN-1 modulates autophagy.

Vps34-IN-1 is an inhibitor of Vps34 extracted from patent WO2012085815A1, compound example 16a, with an IC50 of 4 nM. Vps34-IN-1 modulates autophagy.

Specifications

Chemical Name or Material Vps34-IN-1
CAS 1383716-33-3
Color White
Purity Grade Notes Research
Molecular Formula C21H24ClN7O
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (72.79 mM)
SMILES CC(C)(O)CNC1=NC=C(C2=CC=NC(NC3=CC=NC(Cl)=C3)=N2)C(CC4CC4)=N1
Molecular Weight (g/mol) 425.91
Formula Weight 425.91
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Vps34-IN-1, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.