missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
VPS25 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06483F
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
VPS25 Polyclonal antibody specifically detects VPS25 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| VPS25 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| Dermal papilla-derived protein 9, DERP9, DERP9hVps25, EAP20, EAP20FAP20, ELL-associated protein of 20 kDa, ESCRT-II complex subunit VPS25, FAP20, MGC10540, vacuolar protein sorting 25 (yeast), vacuolar protein sorting 25 homolog (S. cerevisiae), vacuolar protein-sorting-associated protein 25 | |
| Produced in rabbits immunized with E. coli-derived Human VPS25 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 84313 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion