Beskrivelse
VPREB3 Polyclonal antibody specifically detects VPREB3 in Human samples. It is validated for ELISA,Western Blot
Tekniske data
Tekniske data
| Antigen | VPREB3 |
| Applikationer | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | mFluor Violet 610 SE |
| Formulering | 50mM Sodium Borate |
| Gene Alias | N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3 |
| Værtsarter | Rabbit |
| Immunogen | pre-B lymphocyte 3. (Uniprot# Q9UKI3) |
| Oprensningsmetode | Immunogen affinity purified |
| Mængde | 0.1 mL |
| Vis mere |