Få mere at vide
Vortioxetine, TRC
Artikelnummer. 30005748
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005748 100 mg
30005363 5 mg
30004462 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005748 Leverandør Toronto Research Chemicals Leverandørnr. TRCV760100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 508233-74-7
Molekylær formel C18 H22 N2 S
Molekylvægt (g/mol) 298.45
Synonym Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine, Brintellix, Lu AA 21004, Trintellix, Vortioxetine
IUPAC navn 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine
SMIL Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1
Tekniske data
Kemisk navn eller materiale Vortioxetine
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C18H22N2S/c1-14-7-8-17(15(2)13-14)21-18-6-4-3-5-16(18)20-11-9-19-10-12-20/h3-8,13,19H,9-12H2,1-2H3
Formel vægt 298.1504
Procent renhed >95%