Få mere at vide
Vonoprazan Dimer Impurity, TRC
Artikelnummer. 30454651
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454651 50 mg
30453263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30454651 Leverandør Toronto Research Chemicals Leverandørnr. TRCV76700550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vonoprazan Dimer Impurity
CAS 2250243-23-1
Molekylær formel C33H27F2N5O4S2
Mængde 50 mg
Synonym 1H-Pyrrole-3-methanamine','5-(2-fluorophenyl)-N-[[5-(2-fluorophenyl)-1-(3-pyridinylsulfonyl)-1H-pyrrol-3-yl]methyl]-N-methyl-1-(3-pyridinylsulfonyl)- (ACI)
InChI formel InChI=1S/C33H27F2N5O4S2/c1-38(20-24-16-32(28-10-2-4-12-30(28)34)39(22-24)45(41,42)26-8-6-14-36-18-26)21-25-17-33(29-11-3-5-13-31(29)35)40(23-25)46(43,44)27-9-7-15-37-19-27/h2-19,22-23H,20-21H2,1H3
SMIL O=S(=O)(C=1C=NC=CC1)N2C=C(C=C2C=3C=CC=CC3F)CN(C)CC=4C=C(C=5C=CC=CC5F)N(C4)S(=O)(=O)C=6C=NC=CC6
IUPAC navn 1-(5-(2-fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)-N-((5-(2-fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)methyl)-N-methylmethanamine
Molekylvægt (g/mol) 659.73
Formel vægt 659.15