missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Von Willebrand Factor Antibody (Ajinomoto patent anti-vWF) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28401JF525
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Von Willebrand Factor | |
| Monoclonal | |
| Janelia Fluor 525 | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| vWF | |
| 0.1 mL | |
| Cancer | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Ajinomoto patent anti-vWF | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion