Få mere at vide
Volinanserin-d4 Hydrochloride Salt, TRC
Artikelnummer. 30453721
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30453721 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453721 Leverandør Toronto Research Chemicals Leverandørnr. TRCV75500210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Volinanserin-d4 Hydrochloride Salt
CAS 1217617-73-6
Molekylær formel C22H25D4ClFNO3
Mængde 10 mg
Synonym (αR)-α-(2,3-Dimethoxyphenyl)-1-[2-(4-fluorophenyl)ethyl-d4]-4-piperidinemethanol Hydrochloride, (+)-α-(2,3-Dimethoxyphenyl)-1-[2-(4-fluorophenyl)ethyl-d4]-4-piperidinemethanol Hydrochloride, (+)-MDL 100907-d4, M 100907-d4, MDL 100907-d4, R-MDL 100907-d4
InChI formel InChI=1S/C22H28FNO3.ClH/c1-26-20-5-3-4-19(22(20)27-2)21(25)17-11-14-24(15-12-17)13-10-16-6-8-18(23)9-7-16;/h3-9,17,21,25H,10-15H2,1-2H3;1H/t21-;/m1./s1/i10D2,13D2;
SMIL FC(C=C1)=CC=C1C([2H])([2H])C([2H])([2H])N(CC2)CCC2[C@@H](O)C3=C(OC)C(OC)=CC=C3.Cl
Molekylvægt (g/mol) 413.95
Formel vægt 413.2071