Learn More
Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipette with Works Certificate
Individually marked with the time of flow to transfer. Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipettes with Works Certificate are guaranteed to give both volumes and flow times within Class A limits.
Brand: Poulten Graf™ US371/WAC/J/5
This item is not returnable.
View return policy
Description
- Individually numbered and meets the accuracies of USP/ISO/BS and DIN standards
- Amber stain graduation
- Calibrated for delivery
Specifications
Certificate of calibration/Satisfying USP/ISO/BS and DIN accuracies | |
Soda Lime Glass | |
5 Pack | |
50 mL |
A | |
Yes | |
0.05 mL | |
Bulb Pipet |
Product Content Correction
Product Title
Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipette with Works Certificate > Capacity: 50mL
Spot an opportunity for improvement?Share a Content Correction