Få mere at vide
Poulten Graf™ Volac™ Class A Borosilicate Glass Graduated Cylinders with Certificate
Tekniske data
Tekniske data
| Materiale | Borosilicate Glass |
| Kapacitet (metrisk) | 10 mL |
| Inkluderer | Single Certificate |
| Skala | White Ceramic Glazed Ink Scale |
| Gradueringer | 0.2 mL |
| Autoklaverbar | Autoclavable |
| Kalibrering | To Contain |
| Base Type | Hexagonal |
| Skala farve | Blue |
| Form | Round |
| Vis mere |