missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDLR Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Bio-Techne NBP2-99696H
Informations supplémentaires : Weight : 0.00970kg
Description
VLDLR Polyclonal specifically detects VLDLR in Human samples. It is validated for ELISA.Spécification
VLDLR | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
HRP | |
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
Produced in rabbits immunized with purified, recombinant VLDLR (NP_003374.3; Met 1-Ser 797). | |
0.1 mL | |
Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
7436 | |
Store at 4°C in the dark. | |
IgG |