Få mere at vide
Sartorius Vivaspin™ 20
Shop alle Sartorius produkter
nr. pr. pakke:
12 Pk.
48 Pk.
Pakningsstørrelse:
Pakke med 12
Beskrivelse
- CE certified according to EU 98/79/EC IVDD
- Easy-to-use centrifugal or solvent absorption devices
- No risk of sample cross-contamination
- Fast concentration with minimal membrane fouling
- High sample recoveries for increased sensitivity
- Printed graduations on the concentrator body provide a convenient indication of retentate volume and simple, direct recovery from the integral dead-stop pocket is possible with standard pipettes.
- Typical Applications:
- Concentration of patient samples prior to electrophoresis or immuno-electrophoresis
- Concentration of urine, cerebrospinal fluid, or plasma to detect disease markers such as Bence Jones Protein
- Deproteinization of blood samples to aid detection of disease metabolites
Tekniske data
Tekniske data
| Materiale | Polycarbonate |
| Materiale (membran) | PES |
| MWCO | 10 kDa |
| Til brug med (applikation) | For in vitro diagnostics (IVD) applications. |
| nr. pr. pakke | 48 Pk. |