Få mere at vide
Vitamin D5, TRC
Artikelnummer. 30451622
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451622 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451622 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6762201MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vitamin D5
CAS 71761-06-3
Molekylær formel C29 H48 O
Mængde 1 mg
Synonym 24R-Methylcalciol, (3β,5Z,7E)-9,10-Secostigmasta-5,7,10(19)-trien-3-ol, (1S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R,4R)-4-Ethyl-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexanol, (S,E)-3-((Z)-2-((1R,3aS,7aR)-1-((2R,5R)-5-Ethyl-6-methylheptan-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol
InChI formel InChI=1S/C29H48O/c1-7-23(20(2)3)12-10-22(5)27-16-17-28-24(9-8-18-29(27,28)6)13-14-25-19-26(30)15-11-21(25)4/h13-14,20,22-23,26-28,30H,4,7-12,15-19H2,1-3,5-6H3/b24-13+,25-14-/t22-,23-,26+,27-,28+,29-/m1/s1
SMIL CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C)C(C)C
IUPAC navn (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molekylvægt (g/mol) 412.691
Formel vægt 412.371