Få mere at vide
Vitamin D3 Hemisuccinate (~90%), TRC
Artikelnummer. 30450104
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30450104 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450104 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6760852.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vitamin D3 Hemisuccinate
CAS 64889-68-5
Molekylær formel C31H48O4
Mængde 2.5 mg
Synonym Cholecalciferol 3-Hemisuccinate, Cholecalciferol Hemisuccinate, Vitamin D3 Hemisuccinate, 1-[(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexyl] Ester Butanedioic Acid, (3β,5Z,7E)-9,10-Secocholesta-5,7,10(19)-trien-3-ol Hydrogen Butanedioate
InChI formel InChI=1S/C31H48O4/c1-21(2)8-6-9-23(4)27-15-16-28-24(10-7-19-31(27,28)5)12-13-25-20-26(14-11-22(25)3)35-30(34)18-17-29(32)33/h12-13,21,23,26-28H,3,6-11,14-20H2,1-2,4-5H3,(H,32,33)/b24-12+,25-13-/t23-,26+,27-,28+,31-/m1/s1
SMIL [H][C@]1([C@H](C)CCCC(C)C)CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](OC(CCC(O)=O)=O)CC3)/C3=C
Molekylvægt (g/mol) 484.71
Formel vægt 484.3553