Få mere at vide
Vitamin B1 Monophosphate Chloride, TRC
Artikelnummer. 17974265
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17974265 1 g -
17963845 2.5 g 2.5g
17901465 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17974265 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6760151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 532-40-1
Molekylær formel C12 H18 N4 O4 P S . Cl
Molekylvægt (g/mol) 380.79
Synonym Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(phosphonooxy)ethyl]-, chloride (1:1), Phosphoric acid, thiamine ester (6CI), Thiamine, dihydrogen phosphate (ester) (8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(phosphonooxy)ethyl]-, chloride (9CI), Extraneurina, Fosbione, Monophosphothiamine, Neurifosforil, Novaneurina, Novobeta, Phosbitamin, Phosphothiamin, Phosphothiamine, Sineurina, Superneurina, TMP, TMP (thiamin ester), Thiamin monophosphate, Thiamine monophosphate, Thiamine monophosphate chloride, Thiamine phosphate, Thiamine phosphoric acid ester chloride, Thiamine-P, Vitamin B1 monophosphate, Vitamin B1 monophosphate chloride, Vitamin B1 phosphate, Thiamine Monophosphate Chloride
IUPAC navn 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl dihydrogen phosphate;chloride
SMIL [Cl-].Cc1ncc(C[n+]2csc(CCOP(=O)(O)O)c2C)c(N)n1
Tekniske data
Kemisk navn eller materiale Vitamin B1 Monophosphate Chloride
Mængde 1 g
InChI formel InChI=1S/C12H17N4O4PS.ClH/c1-8-11(3-4-20-21(17,18)19)22-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H3-,13,14,15,17,18,19);1H
Formel vægt 380.0475