missing translation for 'onlineSavingsMsg'
Få mere at vide

Vitamin B1 Monophosphate Chloride, TRC

Artikelnummer. 17963845
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17963845 2.5 g 2.5g
17901465 5 g -
17974265 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17963845 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6760152.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 532-40-1
Molekylær formel C12 H18 N4 O4 P S . Cl
Molekylvægt (g/mol) 380.79
Synonym Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(phosphonooxy)ethyl]-, chloride (1:1), Phosphoric acid, thiamine ester (6CI), Thiamine, dihydrogen phosphate (ester) (8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(phosphonooxy)ethyl]-, chloride (9CI), Extraneurina, Fosbione, Monophosphothiamine, Neurifosforil, Novaneurina, Novobeta, Phosbitamin, Phosphothiamin, Phosphothiamine, Sineurina, Superneurina, TMP, TMP (thiamin ester), Thiamin monophosphate, Thiamine monophosphate, Thiamine monophosphate chloride, Thiamine phosphate, Thiamine phosphoric acid ester chloride, Thiamine-P, Vitamin B1 monophosphate, Vitamin B1 monophosphate chloride, Vitamin B1 phosphate, Thiamine Monophosphate Chloride
IUPAC navn 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl dihydrogen phosphate;chloride
SMIL [Cl-].Cc1ncc(C[n+]2csc(CCOP(=O)(O)O)c2C)c(N)n1

Tekniske data

Kemisk navn eller materiale Vitamin B1 Monophosphate Chloride
Mængde 2.5 g
InChI formel InChI=1S/C12H17N4O4PS.ClH/c1-8-11(3-4-20-21(17,18)19)22-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H3-,13,14,15,17,18,19);1H
Formel vægt 380.0475
Produkttitel
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.