Få mere at vide
Vitamin A-d5 Acetate, TRC
Artikelnummer. 30456716
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456716 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456716 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6760021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vitamin A-d5 Acetate
CAS 1217219-62-9
Molekylær formel C22H27D5O2
Mængde 1 mg
Synonym (all-E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl-d5)-2-4,6,8-nonatetraen- 1-ol Acetate, Retinyl-d5 Acetate, Vitamin A1-d5 Acetate, all-trans-Retinol-d5 Acetate
InChI formel InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/i3D3,11D2
SMIL [2H]C1([2H])CCC(C)(C)C(/C=C/C(C)=C/C=C/C(C)=C/COC(C)=O)=C1C([2H])([2H])[2H]
IUPAC navn [9-[3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenyl] acetate
Molekylvægt (g/mol) 333.52
Formel vægt 333.2716