Learn More
Macherey-Nagel™ Visocolor™ HE Titration Test Kit Spare Syringe
Replace the titration syringe from test kits. Macherey-Nagel™ Visocolor™ HE Titration Test Kit Spare Syringes are used with HE tests to determine even the lowest limiting values. The gradation marks reveal the level of the tested parameter that is equivalent to the amount of titrant used.
Brand: Macherey-Nagel™ 915405
Additional Details : Weight : 0.50000kg
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the reagents, as well as by compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests. Each test kit is economically priced and provides convenient handling.
- High sensitive volumetric analysis via visual evaluation
- Narrow syringe with gradation marks for easy, accurate readings right on the barrel
- Gradations show equivalent analyte level based on titrant required for color change
- Sharper color change due to separated dropping reagents
- Refill packs and replacement titration syringes available
- Single reagent bottles with clear dosing instructions
- Delivered in a robust box with plastic inlay and easy-to-understand instructions
Specifications
Calcium (Ca2+) | |
Total hardness determination | |
Graduated Syringe | |
Titration syringe, dropping tips | |
1 Each | |
Colorimetric titration |
Clear barrel, black graduation marks, white plunger with black o-ring | |
Visocolor™ HE Test Kit for Total Hardness HA 20 (915 005) | |
0.5 °d or 0.1mmol/L gradation mark | |
200 | |
0.5 to 20.0 °d or 0.1 to 3.6mmol/L Ca2+ | |
Titration syringe |