Learn More
Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit Refill Pack
Replace consumed chemicals from test kits. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit Refill Packs are reagents used with HE tests to determine even the lowest limiting values. The kits use longer measuring tubes and larger sample volumes for the highest sensitivity and accuracy.
Brand: Macherey-Nagel™ 920174
Additional Details : Weight : 0.50000kg
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.
- Visual evaluation
- Environment-friendly, without toxic reagents
- Economically priced
- Convenient handling
- Highest accuracy due to extremely narrow gradation
- Highest sensitivity down to 0.002mg/L due to longer measuring tubes
- Reagent bottles with clear dosing instructions
- Compensation of turbidities and colors
- Refill packs available
Specifications
pH | |
Highly sensitive | |
HE Colorimetric Test Kit, pH 4 - 10 (mfr. no. 920 074) | |
4.0 - 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 - 10.0 pH | |
500 | |
4.0 to 10.0 pH | |
30 sec. |
Color change: Orange - Yellow - Green - Brown | |
pH determination | |
Liquid Reagent | |
Reagent bottles | |
1 Each | |
Mixed indicator | |
Reagent set |