Learn More
Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit
Provides the highest sensitivity and accuracy in visual analytics. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kits determine even the lowest limiting values using longer measuring tubes and larger sample volumes. Includes all reagents and testing equipment/supplies.
Brand:  Macherey-Nagel™ 920063
Additional Details : Weight : 20.00000kg
 View more versions of this product
Product Code. 10421151
  • 708.00 DKK / 1 test
Estimated Shipment
29-03-2021
Add to basket
Description
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.
  • Visual evaluation
  • Environment-friendly, without toxic reagents
  • Economically priced
  • Convenient handling
  • Highest accuracy due to extremely narrow gradation
  • Highest sensitivity down to 0.002mg/L due to longer measuring tubes
  • Reagent bottles with clear dosing instructions
  • Compensation of turbidities and colors
  • Refill packs available
Specifications
Specifications
Nitrite (NO2-)
Highly sensitive
HE Colorimetric Test Kit
Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk)
1 Each
Sulfanilic acid / 1-naphthylamine
Water analysis test kit
Color change: Yellow to Bright Pink
Nitrite determination
0.0 - 0.005 - 0.010 - 0.015 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10mg/L NO2-
150
0 to 0.10mg/L
10 min.