Learn More
Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit
Provides the highest sensitivity and accuracy in visual analytics. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kits determine even the lowest limiting values using longer measuring tubes and larger sample volumes. Includes all reagents and testing equipment/supplies.
Brand: Macherey-Nagel™ 920063
Additional Details : Weight : 20.00000kg
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.
- Visual evaluation
- Environment-friendly, without toxic reagents
- Economically priced
- Convenient handling
- Highest accuracy due to extremely narrow gradation
- Highest sensitivity down to 0.002mg/L due to longer measuring tubes
- Reagent bottles with clear dosing instructions
- Compensation of turbidities and colors
- Refill packs available
Specifications
Nitrite (NO2-) | |
Highly sensitive | |
HE Colorimetric Test Kit | |
Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk) | |
1 Each | |
Sulfanilic acid / 1-naphthylamine | |
Water analysis test kit |
Color change: Yellow to Bright Pink | |
Nitrite determination | |
0.0 - 0.005 - 0.010 - 0.015 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10mg/L NO2- | |
150 | |
0 to 0.10mg/L | |
10 min. |