missing translation for 'onlineSavingsMsg'
Learn More

Viscosity Reference Standard Low Temperature Type N480B UKAS ISO 17025 / ISO 17034 Certified, 500 mL, Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

Brand:  Paragon Scientific N480B

Additional Details : Weight : 0.50000kg


Product Code. 15588874

  • 695.00 DKK / 500mL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL, available also in 5L. Please contact sales@paragon-sci.com for other volumes
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • low temperature viscosity standard

    Signal Word
    • Danger

    Hazard Category
    • Aspiration toxicity Category 1

    Hazard Statement
    • H304-May be fatal if swallowed and enters airways.

    Precautionary Statement
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P331-Do NOT induce vomiting.

    Supplemental information
    • MIXTURE LIST-Contains : baseoil - unspecified

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Nominal values quoted. Certified Values will vary from batch to batch.