Learn More
Poulten Selfe™ Viscometer Holder
Product Code. 13121103
Change view
Click to view available options
Size:
Cannon-Fenske Size 25 to 500
Suspended Level Sizes 1 to 4
Suspended Level Sizes 4a to 5
U-tube Reverse Flow Sizes 1 to 9
U-tube Size A to F
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
13121103 Cannon-Fenske Size 25 to 500 Each
13191093 Suspended Level Sizes 1 to 4 Each
13101103 Suspended Level Sizes 4a to 5 Each
13181093 U-tube Reverse Flow Sizes 1 to 9 Each
13161093 U-tube Size A to F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13121103 Supplier Poulten Selfe™ Supplier No. 1833/A1
  • ONLINE EXCLUSIVE PRICE
    1140.00 DKK
    1077.11 DKK / Pakke med 1
    Spare 62.89 DKK
    5% Slukket
 Restricted Item
Forventet forsendelse: 02-02-2027
Specifications
Product Type Viscometer Holder
For Use With (Equipment) Cannon-Fenske Routine 25 to 500
Diameter (Metric) Opening 50 mm
Size Cannon-Fenske Size 25 to 500