Få mere at vide
Thermo Scientific™ Virtuoso™ Vial Identification System
Artikelnummer. 15201918
Skift visning
Pakningsstørrelse:
Hver
Artikelnummer. Pakningsstørrelse
15201918 Hver
1 options
Artikelnummer. 15201918 Leverandør Thermo Scientific™ Leverandørnr. 60180VT100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Eliminate errors from manual vial labeling. This advanced labeling-information device ensures accurate sample identification and increases productivity.
The Thermo Scientific™ Virtuoso™ Vial Identification System is an error-free option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow. With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity. Use the system’s touch screen and network capabilities to transfer text, graphics, barcodes, and logos directly onto your vials.
Facilitates Your Labeling Requirements
  • Fully loaded with standard or fully user-defined templates
Increases Productivity
  • Eliminate error-prone manual vial labeling
  • Label a vial in just 5 seconds
  • Improve labeling efficiencies by downloading sample lists
  • Simplify vial sample identification
Produces High-Quality Labeling
  • Resists common laboratory solvents, blurring, and smudging
Provides an Advanced Labeling and information System in One Device
  • Designed by experienced analytical scientists
TRUSTED_SUSTAINABILITY
Tekniske data
Produktlinje Virtuoso
Produkttype Vial Identification System
Certificeringer/compliance Pollution Level 2
Højde (engelsk) 11.61 in.
Højde (metrisk) 295 mm
Længde (engelsk) 7.87 in.
Længde (metrisk) 200 mm
Strømforsyning Universal power supply 100-240V AC
Bredde (metrisk) 140 mm