Få mere at vide
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Shop alle Ansell produkter
Størrelse:
7
8
9
10
11
Pakningsstørrelse:
50 par
Beskrivelse
- Reusable
- Anti-static according to EN1149
- With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
- Significantly lower tendency to shed than traditional cotton flock gloves
- Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
Tekniske data
Tekniske data
| Materiale bagpå | Nitrile |
| Manchet materiale | Nitrile |
| Produkttype | Immersion Gloves |
| Størrelse | 7 |
| Stil | 79-700 |
| Tykkelse (metrisk) | 0.2 mm |
| Handske manchet farve | Blue |
| Handske manchet stil | Straight |
| Liner materiale | Aquadri |
| Mængde | 1 Pair |
| Vis mere |