Få mere at vide
Vioxx-d5 (Major), TRC
Artikelnummer. 30453204
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30453204 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453204 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6700021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vioxx-d5
CAS 544684-93-7
Molekylær formel C17 2H5 H9 O4 S
Mængde 1 mg
Synonym 4-[4-(Methylsulfonyl)phenyl]-3-(phenyl-d5)-2(5H)-furanone, 3-(4-Methanesulfonylphenyl)-2-(phenyl-d5)-2-buten-4-olide, 3-(Phenyl-d5)-4-[4-(Methylsulfonyl)phenyl]-2(5H)-furanone, MK 0966-d5, MK 966-d5, Rhuma-cure-d5, Rofecoxib-d5
InChI formel InChI=1S/C17H14O4S/c1-22(19,20)14-9-7-12(8-10-14)15-11-21-17(18)16(15)13-5-3-2-4-6-13/h2-10H,11H2,1H3/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C2=C(COC2=O)c3ccc(cc3)S(=O)(=O)C)c([2H])c1[2H]
IUPAC navn 3-(4-methylsulfonylphenyl)-4-(2,3,4,5,6-pentadeuteriophenyl)-2H-furan-5-one
Molekylvægt (g/mol) 319.387
Formel vægt 319.093