missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Vinyltrimethoxysilane, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trimethoxyvinylsilane |
| CAS | 2768-02-7 |
| Molekylær formel | C5 H12 O3 Si |
| Mængde | 25 g |
| Synonym | Silane','ethenyltrimethoxy-, Silane','trimethoxyvinyl- (6CI,7CI,8CI), Ethenyltrimethoxysilane, (Trimethoxysilyl)ethene, A 101, A 101 (silane), A 171, AC 71, CV 4917, DB 171, DL 171, Dow Corning 6300, Dynasylan Si 108, Dynasylan Silfin, Dynasylan Silfin 24, Dynasylan VTMO, EMI 1833, GX 171, Geniosil X 10GS, Geniosil XL 10, KBM 1003, KBM 3100, KH 171, KH 921, KHA 171, LMD 171, LS 815, LT 171, NUC Silicone A 171, NUC-Y 9818, Penta 1002, Q 9-6300, S 210, SB 6301, SCA 1603, SG-Si 171, SH 6300, SIV 9220.0, SL 815, SZ 6300, Sigma T 5051, Sila-Ace 210, Sila-Ace S 210, Silane A 171, Silane V Trimethoxy, Silaplane S 210, Silfin 13, Silfin 22, Silfin 24, Silfin 59, Silopren TP 3625, Silox VS 911, Silquest A 171, TSL 8310, Trimethoxyvinylsilane, U 611, UN 1993, UP 171, V 0042, V 06, V 06 (coupling agent), V 4917, VTMO, VTMS, VTS-M, Vinyltrimethoxysilane, WD 21, XL 10, XL 10 (silane), Xiameter OFS 6300, Y 4302, YDH 171, YGO 1202, Z 6300 |
| InChI formel | InChI=1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3 |
| SMIL | CO[Si](OC)(OC)C=C |
| IUPAC navn | ethenyl(trimethoxy)silane |
| Molekylvægt (g/mol) | 148.2325 |
| Formel vægt | 148.0556 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?