Få mere at vide
Vinyltrimethoxysilane, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trimethoxyvinylsilane |
| CAS | 2768-02-7 |
| Molekylær formel | C5 H12 O3 Si |
| Mængde | 50 g |
| Synonym | Silane','ethenyltrimethoxy-, Silane','trimethoxyvinyl- (6CI,7CI,8CI), Ethenyltrimethoxysilane, (Trimethoxysilyl)ethene, A 101, A 101 (silane), A 171, AC 71, CV 4917, DB 171, DL 171, Dow Corning 6300, Dynasylan Si 108, Dynasylan Silfin, Dynasylan Silfin 24, Dynasylan VTMO, EMI 1833, GX 171, Geniosil X 10GS, Geniosil XL 10, KBM 1003, KBM 3100, KH 171, KH 921, KHA 171, LMD 171, LS 815, LT 171, NUC Silicone A 171, NUC-Y 9818, Penta 1002, Q 9-6300, S 210, SB 6301, SCA 1603, SG-Si 171, SH 6300, SIV 9220.0, SL 815, SZ 6300, Sigma T 5051, Sila-Ace 210, Sila-Ace S 210, Silane A 171, Silane V Trimethoxy, Silaplane S 210, Silfin 13, Silfin 22, Silfin 24, Silfin 59, Silopren TP 3625, Silox VS 911, Silquest A 171, TSL 8310, Trimethoxyvinylsilane, U 611, UN 1993, UP 171, V 0042, V 06, V 06 (coupling agent), V 4917, VTMO, VTMS, VTS-M, Vinyltrimethoxysilane, WD 21, XL 10, XL 10 (silane), Xiameter OFS 6300, Y 4302, YDH 171, YGO 1202, Z 6300 |
| InChI formel | InChI=1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3 |
| SMIL | CO[Si](OC)(OC)C=C |
| IUPAC navn | ethenyl(trimethoxy)silane |
| Molekylvægt (g/mol) | 148.2325 |
| Formel vægt | 148.0556 |