Få mere at vide
Vinpocetine EP Impurity B (Vincamine EP Impurity D), Mikromol™
Beskrivelse
Vinpocetine EP Impurity B (Vincamine EP Impurity D)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Apovincamine |
| CAS | 4880-92-6 |
| Analyt- eller komponentnavne | Apovincamine |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C21 H24 N2 O2 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Methyl (13aS,13bS)-13a-Ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate (Apovincamine), Vinpocetine Imp. B (EP), Vinpocetine USP Related Compound B, Vinpocetine USP RC B, Methyl (13aS,13bS)-13a-Ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate, Apovincamine, Vinpocetine Related Compound B, Vincamine Impurity D, Vinpocetine Impurity B |
| InChI formel | InChI=1S/C21H24N2O2/c1-3-21-10-6-11-22-12-9-15-14-7-4-5-8-16(14)23(18(15)19(21)22)17(13-21)20(24)25-2/h4-5,7-8,13,19H,3,6,9-12H2,1-2H3/t19-,21+/m1/s1 |
| SMIL | CC[C@@]12CCCN3CCc4c([C@H]13)n(C(=C2)C(=O)OC)c5ccccc45 |
| IUPAC navn | Methyl (13aS,13bS)-13a-Ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate |
| Vis mere |