Få mere at vide
Vindoline, TRC
Artikelnummer. 30137825
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137825 5 mg
30134751 50 mg
30007185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137825 Leverandør Toronto Research Chemicals Leverandørnr. TRCV4165005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2182-14-1
Molekylær formel C25 H32 N2 O6
Molekylvægt (g/mol) 456.53
Synonym Aspidospermidine-3-carboxylic acid, 4-(acetyloxy)-6,7-didehydro-3-hydroxy-16-methoxy-1-methyl-, methyl ester, (2β,3β,4β,5α,12R,19α)- (9CI), (-)-Vindoline, Vindoline
IUPAC navn methyl (1R,9R,10S,11R,12R,19R)-11-acetyloxy-12-ethyl-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.01,9.02,7.016,19]nonadeca-2(7),3,5,13-tetraene-10-carboxylate
SMIL CC[C@@]12C=CCN3CC[C@@]4([C@H]13)[C@@H](N(C)c5cc(OC)ccc45)[C@](O)([C@@H]2OC(=O)C)C(=O)OC
Tekniske data
Kemisk navn eller materiale Vindoline
Mængde 5 mg
InChI formel InChI=1S/C25H32N2O6/c1-6-23-10-7-12-27-13-11-24(19(23)27)17-9-8-16(31-4)14-18(17)26(3)20(24)25(30,22(29)32-5)21(23)33-15(2)28/h7-10,14,19-21,30H,6,11-13H2,1-5H3/t19-,20+,21+,23+,24+,25-/m0/s1
Formel vægt 456.226