missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Vinculin Antibody (VCL/2575), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79942DL594
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Vinculin Monoclonal specifically detects Vinculin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| Vinculin | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| VCL | |
| Recombinant fragment (around aa 174-322) of human Vinculin (VCL) protein (exact sequence is proprietary) (Uniprot: P18206) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 7414 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| VCL/2575 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion