Få mere at vide
Vincristine N-Oxide, TRC
Artikelnummer. 30004359
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004359 2.5 mg
30006978 25 mg
30012376 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004359 Leverandør Toronto Research Chemicals Leverandørnr. TRCV3141002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 947527-73-3
Molekylær formel C46 H56 N4 O11
Molekylvægt (g/mol) 840.96
Synonym 22-Oxo-vincaleukoblastine 6'-Oxide
SMIL CC[C@]1(O)C[C@H]2C[C@](C(=O)OC)(c3cc4c(cc3OC)N(C=O)[C@H]5[C@](O)([C@H](OC(=O)C)[C@]6(CC)C=CCN7CC[C@]45[C@H]67)C(=O)OC)c8[nH]c9ccccc9c8CCN(=O)(C2)C1
Tekniske data
Kemisk navn eller materiale Vincristine N-Oxide
Mængde 2.5 mg
InChI formel InChI=1S/C46H56N4O11/c1-7-42(55)22-28-23-45(40(53)59-5,36-30(14-19-50(57,24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)58-4)49(26-51)38-44(31)16-18-48-17-11-15-43(8-2,37(44)48)39(61-27(3)52)46(38,56)41(54)60-6/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3/t28-,37-,38+,39+,42-,43+,44+,45-,46-,50-/m0/s1
Formel vægt 840.3946