Få mere at vide
Vincristine-d3 Sulfate, TRC
Artikelnummer. 30452219
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30452219 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452219 Leverandør Toronto Research Chemicals Leverandørnr. TRCV3142531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vincristine-d3 Sulfate
CAS 1246817-10-6
Molekylær formel C46H55D3N4O14S
Mængde 1 mg
Synonym 22-Oxovincaleukoblastine-d3 Sulfate, Leurocristine-d3 Sulfate, Kyocristine-d3, Lilly 37231-d3, NSC 67574-d3, Novopharm-d3, Oncovin-d3, Onkovin-d3, VCR Sulfate-d3, Vincasar PFS-d3, Vincrisul-d3
InChI formel InChI=1S/C46H56N4O10.H2O4S/c1-7-42(55)22-28-23-45(40(53)58-5,36-30(14-18-48(24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)57-4)50(26-51)38-44(31)16-19-49-17-11-15-43(8-2,37(44)49)39(60-27(3)52)46(38,56)41(54)59-6;1-5(2,3)4/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3;(H2,1,2,3,4)/t28-,37+,38-,39-,42+,43-,44-,45+,46+;/m1./s1/i3D3;
SMIL O=S(O)(O)=O.CC[C@@]12[C@@]3([H])[C@]4(CCN3CC=C2)[C@](N(C5=CC(OC)=C([C@@]6(C7=C(CC[N@]8C[C@@](C[C@](O)(C8)CC)([H])C6)C9=CC=CC=C9N7)C(OC)=O)C=C54)C=O)([H])[C@](O)([C@@H]1OC(C([2H])([2H])[2H])=O)C(OC)=O
IUPAC navn methyl (3aR,3a1R,4R,5S,5aR,10bR)-4-(acetoxy-d3)-3a-ethyl-9-((3S,5S,7S,9S)-5-ethyl-5-hydroxy-9-(methoxycarbonyl)-1,4,5,6,7,8,9,10-octahydro-2H-3,7-methano[1]azacycloundecino[5,4-b]indol-9-yl)-6-formyl-5-hydroxy-8-methoxy-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate sulfate
Molekylvægt (g/mol) 926.05
Formel vægt 925.39