missing translation for 'onlineSavingsMsg'
Learn More
Vincamine, MedChemExpress Product Code.: 16438566

Vincamine, MedChemExpress

Product Code. p-200100907
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16438566

Brand: Medchem Express HYB1021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vincamine is a monoterpenoid indole alkaloid extracted from the Madagascar periwinkle.

Vincamine is a peripheral vasodilator and exerts a selective vasoregulator action on the brain microcapilar circulation. Vincamine is a GPR40 agonist and acts as a β-cell protector by ameliorating β-cell dysfunction and promoting glucose-stimulated insulin secretion (GSIS). Vincamine improves glucose homeostasis in vivo, and has the potential for the type 2 diabetes mellitus (T2DM) research.

Specifications

Chemical Name or Material Vincamine
CAS 1617-90-9
Color White
Purity Grade Notes Research
Molecular Formula C21H26N2O3
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (70.53 mM; Need ultrasonic)
SMILES O=C(OC)[C@@]1(O)C[C@@]2(CC)[C@@]3([H])C(N1C4=CC=CC=C54)=C5CCN3CCC2
Molecular Weight (g/mol) 354.44
Formula Weight 354.44
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Vincamine, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.