Få mere at vide
Viloxazine Hydrochloride, LoGiCal
Artikelnummer. 30458035
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458035 10 mg
1 options
Artikelnummer. 30458035 Leverandør LoGiCal Leverandørnr. LGCFOR3361.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Viloxazine Hydrochloride
CAS 35604-67-2
Analyt- eller komponentnavne Viloxazine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H19 N O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym Morpholine, 2-[(2-ethoxyphenoxy)methyl]-, hydrochloride (1:1), Morpholine, 2-[(2-ethoxyphenoxy)methyl]-, hydrochloride (9CI), 2-[(2-Ethoxyphenoxy)methyl]morpholinium chloride, Catatrol, ICI 58834, Vicilan, Viloxazin hydrochloride, Viloxazine hydrochloride, Vivalan, Vivarint, Vivilan
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H
Løsningstype Neat
SMIL Cl.CCOc1ccccc1OCC2CNCCO2
IUPAC navn 2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 273.76
Mængde 10 mg
Formel vægt 273.1132 g/mol