Få mere at vide
Tocris Bioscience™ Vilazodone hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 6202/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Vilazodone hydrochloride is a potent 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SSRI) (IC50 values are 0.2 and 0.5 nM respectively). Antidepressant.Tekniske data
| Potent 5-HT1A Partial Agonist and SSRI, Antidepressant | |
| 163521-08-2 | |
| 477.99g/mol | |
| 5-HT1A Receptor Antagonists |
| 5-HT1A | |
| C26H27N5O2.HCl | |
| Store at -20°C | |
| 10 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only