Få mere at vide
Vilazodone.HCl, Lipomed
Artikelnummer. 30458702
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30458702 100 mg
1 options
Artikelnummer. 30458702 Leverandør Lipomed Leverandørnr. LPMVZD1891HC100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Vilazodone.HCl
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vilazodone HCl
CAS 163521-08-2
Analyt- eller komponentnavne Vilazodone HCl
Forsendelsestilstand Room Temperature
Molekylær formel C26 H27 N5 O2 . Cl H
Synonym 2-Benzofurancarboxamide, 5-[4-[4-(5-cyano-1 H-indol-3-yl)butyl]-1-piperazinyl]-, hydrochloride (1:1), 2-Benzofurancarboxamide, 5-[4-[4-(5-cyano-1 H-indol-3-yl)butyl]-1-piperazinyl]-, monohydrochloride (9 CI), EMD 68843, SB 659746 A, Viibryd, Vilazodone hydrochloride
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMIL Cl.NC(=O)c1oc2ccc(cc2c1)N3CCN(CCCCc4c[nH]c5ccc(cc45)C#N)CC3
IUPAC navn 5-[4-[4-(5-cyano-1 H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide;hydrochloride
Industry Type Pharmaceutical
Molekylvægt (g/mol) 477.99
Mængde 100 mg
Formel vægt 477.1932 g/mol