Få mere at vide
R&D Systems™ Recombinant Human Reelin Protein
Artikelnummer. 18736157
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18736157
Brand: R&D Systems™ 8546MR050
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolicy
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Human Reelin Protein is derived from CHO. The Recombinant Human Reelin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer P78509
Til brug med (applikation) Bioactivity
Gen-id (Entrez) 5649
Molekylvægt (g/mol) Observed M.W.: 155-188kDa, reducing conditions, Theoretical M.W.: 163kDa
Mængde 50 μg
Kilde Chinese Hamster Ovary cell line,CHO-derived human Reelin protein Ser1221-Gln2666,with a C-terminal 6-His tag
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoksin koncentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598
Fælles navn Reelin
Gen symbol RELN
Biologisk aktivitet Measured by its binding ability in a functional ELISA. When Recombinant Human Reelin is coated at 1.5μg/mL (100μL/well), the concentration of Recombinant Human Apolipoprotein E R2/ApoE R2 that produces 50% optimal binding response is typically 50-300ng/mL.
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Udtrykssystem Chinese Hamster Ovary cell line, CHO-derived Ser1221-Gln2666, with a C-terminal 6-His tag
Renhed eller kvalitetsklasse >90%, by SDS-PAGE with Silver Staining