Få mere at vide
VH-298, MedChemExpress
Artikelnummer. 16494486
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494486 100 mg 100mg
16493218 10 mg -
16489766 50 mg 50mg
16460046 2 mg -
16405957 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16494486 Leverandør Medchem Express Leverandørnr. HY100947
MedChemExpress VH-298 is a highly potent inhibitor of the VHL:HIF-α interaction with a Kd value of 80 to 90 nM, used in PROTAC technology.
VH-298 is a highly potent inhibitor of the VHL:HIF-α interaction with a Kd value of 80 to 90 nM, used in PROTAC technology.
Tekniske data
Kemisk navn eller materiale VH-298
CAS 2097381-85-4
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C27H33N5O4S
Mængde 100 mg
Opløselighedsinformation DMSO : ≥ 83.3 mg/mL (159.08 mM)
SMIL O=C([C@H]1N(C([C@H](C(C)(C)C)NC(C2(CC2)C#N)=O)=O)C[C@H](O)C1)NCC3=CC=C(C4=C(C)N=CS4)C=C3
Molekylvægt (g/mol) 523.65
Formel vægt 523.65
Procent renhed 99.78%
Grad Research
Fysisk form Solid