Få mere at vide
Tocris Bioscience™ VH 032, propargyl
Beskrivelse
VH 032, propargyl is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal propargyl ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2S,4R)-1-((S)-3,3-Dimethyl-2-(2-(prop-2-yn-1-yloxy)acetamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2098799-78-9 |
| Mængde | 50 mg |
| Molekylær formel | C27H34N4O5S |
| Formel vægt | 526.65 |
| Produkttype | Alkyne-functionalized Ligand |
| Renhed | 95% |