Få mere at vide
Tocris Bioscience™ VH 032 phenol-alkylC4-amine
Beskrivelse
VH 032 phenol-alkylC4-amine is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was VH 032 phenol - linker 1
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2S,4R)-1-((S)-2-Acetamido-3,3-dimethylbutanoyl)-N-(2-(4-aminobutoxy)-4-(4-methylthiazol-5-yl)benzyl)-4-hydroxypyrrolidine-2-carboxamide dihydrochloride |
| CAS | 2376990-26-8 |
| Synonym | VH 032 phenol - linker 1 |
| Mængde | 25 mg |
| Molekylær formel | C28H41N5O5S.2HCl |
| Formel vægt | 632.64 |
| Produkttype | Functionalized von-Hippel-Lindau (VHL) Ligand |
| Renhed | 95% |