Få mere at vide
Tocris Bioscience™ VH 032 amide-alkylC4-acid
Beskrivelse
VH 032 amide-alkylC4-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus alkylC4 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was VH 032 - linker 5
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 6-((-1-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-6-oxohexanoic acid |
| CAS | 2172819-74-6 |
| Synonym | VH 032 - linker 5 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C28H38N4O6S |
| Formel vægt | 558.69 |
| Produkttype | Functionalized von-Hippel-Lindau (VHL) Ligand |
| Renhed | 95% |