Få mere at vide
VH 032 - linker 3, Tocris Biosciences™
GREENER_CHOICE
PROTACs (Proteolysis Targeting Chimeras)
Brand:  Tocris Bioscience 6465/25
Encompass
Produktkode 15907960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
VH 032 amide-alkylC4-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was VH 032 - linker 3
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
PROTAC Toolbox
2341796-78-7
VH 032 - linker 3
25 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner