Få mere at vide
Vetrabutine Hydrochloride, LoGiCal
Artikelnummer. 30460099
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460099 10 mg
1 options
Artikelnummer. 30460099 Leverandør LoGiCal Leverandørnr. LGCFOR3057.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vetrabutine Hydrochloride
CAS 9-4-5974
Analyt- eller komponentnavne Vetrabutine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H27 N O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym Monzal hydrochloride, Vetrabutin chlorhydrate, Vetrabutine hydrochloride, Benzenebutanamine, alpha-(3,4-dimethoxyphenyl)-N,N-dimethyl-, hydrochloride (1:1), Benzenebutanamine, alpha-(3,4-dimethoxyphenyl)-N,N-dimethyl-, hydrochloride (9CI), Veratrylamine, N,N-dimethyl-alpha-(3-phenylpropyl)-, hydrochloride (6CI,7CI,8CI)
InChI formel InChI=1S/C20H27NO2.ClH/c1-21(2)18(12-8-11-16-9-6-5-7-10-16)17-13-14-19(22-3)20(15-17)23-4;/h5-7,9-10,13-15,18H,8,11-12H2,1-4H3;1H
Løsningstype Neat
SMIL Cl.COc1ccc(cc1OC)C(CCCc2ccccc2)N(C)C
IUPAC navn 1-(3,4-dimethoxyphenyl)-N,N-dimethyl-4-phenylbutan-1-amine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 349.89
Mængde 10 mg
Formel vægt 349.1809 g/mol