Få mere at vide
Verteporfin, TRC
Artikelnummer. 30453813
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453813 10 mg
30450101 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30453813 Leverandør Toronto Research Chemicals Leverandørnr. TRCV12880010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129497-78-5
Molekylær formel C41H42N4O8
Molekylvægt (g/mol) 1437.5884
Synonym 23H,25H-Benzo[b]porphine-9,13-dipropanoic acid, 18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-, monomethyl ester, trans-, BPD-MA, CL 318952, Verteporfin, Visudyne, (4R,4aS)-rel-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-benzo[b]porphine-9,13-dipropanoic acid monomethyl ester
IUPAC navn 3-[rac-(23R,24S)-22,23-bis(methoxycarbonyl)-5-(3-methoxy-3-oxo-propyl)-4,10,15,24-tetramethyl-14-vinyl-25,26,27,28-tetrazahexacyclo[16.6.1.13,6.18,11.113,16.019,24]octacosa-1,3(28),4,6,8,10,12,14,16(26),17,19,21-dodecaen-9-yl]propanoic acid
SMIL COC(=O)CCc1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(CCC(=O)O)c5C)c(C=C)c4C)C6=CC=C([C@H](C(=O)OC)[C@]36C)C(=O)OC.COC(=O)CCc7c(C)c8cc9nc(cc%10[nH]c(cc%11nc(cc7[nH]8)c(CCC(=O)O)c%11C)[C@@]%12(C)[C@@H](C(=O)OC)C(=CC=C%10%12)C(=O)OC)c(C)c9C=C
Tekniske data
Kemisk navn eller materiale Verteporfin
Mængde 10 mg
InChI formel InChI=1S/2C41H42N4O8/c1-9-23-20(2)29-17-34-27-13-10-26(39(49)52-7)38(40(50)53-8)41(27,5)35(45-34)19-30-22(4)24(11-14-36(46)47)32(44-30)18-33-25(12-15-37(48)51-6)21(3)28(43-33)16-31(23)42-29;1-9-23-20(2)29-17-34-27-13-10-26(39(49)52-7)38(40(50)53-8)41(27,5)35(45-34)19-30-22(4)25(12-15-37(48)51-6)33(44-30)18-32-24(11-14-36(46)47)21(3)28(43-32)16-31(23)42-29/h2*9-10,13,16-19,38,43,45H,1,11-12,14-15H2,2-8H3,(H,46,47)/b31-16-,32-18-,34-17-,35-19-;31-16-,33-18-,34-17-,35-19-/t2*38-,41+/m11/s1
Formel vægt 1436.6005