Få mere at vide
Ansell™ VersaTouch™ 92-210 Series Single-Use Blue Nitrile Gloves
Shop alle Ansell produkter
Størrelse:
7
8
9
10
Pakningsstørrelse:
Pakke med 100
Beskrivelse
- Single use gloves
- Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions
- Thinner glove with enhanced fingertip flexibility and improved overall comfort
- Textured fingertips provide good comfort and tactility for sensitive, precise handling
- Latex-free: no risk of Type 1 allergies
- Approved for food contact applications
- Ideal for handling all fatty food
Applications:
- Prepared meals
- Fruit and vegetable processing
- Dairy processing
- Food distribution
- Catering
Tekniske data
Tekniske data
| Udvendig finish | Fully Textured |
| Materiale | Nitrile |
| Størrelse | 8 |
| Tykkelse (engelsk) | 2.9 mil |
| Tykkelse (metrisk) | 0.07 mm |
| Model- | 922108 |
| Serie | 92-210 |