Få mere at vide
Vernolate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Vernolate |
| CAS | 1929-77-7 |
| Molekylær formel | C10 H21 N O S |
| Mængde | 500 mg |
| Synonym | Carbamothioic acid','N,N-dipropyl-','S-propyl ester, Carbamic acid','dipropylthio-','S-propyl ester (6CI,8CI), Carbamothioic acid','dipropyl-','S-propyl ester (9CI), Dipropylthiocarbamic acid S-propyl ester, PPTC, Perbulate, Propyl dipropylthiolcarbamate, R-1607, S-Propyl dipropylthiocarbamate, S-n-Propyl N,N-dipropylthiocarbamate, Vanalate, Vernam, Vernolate |
| InChI formel | InChI=1S/C10H21NOS/c1-4-7-11(8-5-2)10(12)13-9-6-3/h4-9H2,1-3H3 |
| SMIL | CCCSC(=O)N(CCC)CCC |
| IUPAC navn | S-propyl N,N-dipropylcarbamothioate |
| Molekylvægt (g/mol) | 203.34 |
| Formel vægt | 203.1344 |