Få mere at vide
Verlukast-d6, TRC
Artikelnummer. 30456218
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456218 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456218 Leverandør Toronto Research Chemicals Leverandørnr. TRCV1285021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Verlukast-d6
CAS 153698-86-3
Molekylær formel C26 2H6 H21 Cl N2 O3 S2
Mængde 1 mg
InChI formel InChI=1S/C26H27ClN2O3S2/c1-29(2)25(30)12-13-33-17-34-24(16-26(31)32)20-5-3-4-18(14-20)6-10-22-11-8-19-7-9-21(27)15-23(19)28-22/h3-11,14-15,24H,12-13,16-17H2,1-2H3,(H,31,32)/b10-6+/t24-/m1/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C(=O)CCSCS[C@H](CC(=O)O)c1cccc(\C=C\c2ccc3ccc(Cl)cc3n2)c1)C([2H])([2H])[2H]
IUPAC navn (3R)-3-[[3-[bis(trideuteriomethyl)amino]-3-oxopropyl]sulfanylmethylsulfanyl]-3-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]propanoic acid
Molekylvægt (g/mol) 521.12
Formel vægt 520.1528