Learn More
MedChemExpress Verbascoside
Product Code. 16478956
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
16478956 100 mg 100mg
1 options
Product Code. 16478956 Supplier MedChemExpress Supplier No. HYN0021
 Restricted Item
Forventet forsendelse: 30-11-2026
MedChemExpress Verbascoside is isolated from Lantana camara, acts as an ATP-competitive inhibitor of PKC, with an IC50 of 25 μM, and has antitumor, anti-inflammatory and antineuropathic pain activity.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 624.59 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 61276-17-3
Product Type Natural Products
Purity 95.67%
Solubility DMSO : ≥ 100 mg/mL (160.11 mM)
Synonym Acteoside∣ Kusaginin∣ TJC160
Target PKC∣Apoptosis∣Bacterial∣HSV
Quantity 100 mg