Få mere at vide
Verapamil-d6, TRC
Artikelnummer. 30453477
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30453477 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453477 Leverandør Toronto Research Chemicals Leverandørnr. TRCV1249651MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Verapamil-d6
CAS 1185193-17-2
Renhedsgrad noter HPLC
Molekylær formel C27 D6 H32 N2 O4
Mængde 1 mg
Synonym 2-(3,4-Dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)pentanenitrile, Benzeneacetonitrile','α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-[1-(methyl-d3)ethyl-2,2,2-d3]-, (±)-Verapamil-d6, Verapamil-D6 (isopropyl-1,1,1,3,3,3-d6), 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-(isopropyl-1,1,1,3,3,3-d6)-valeronitrile, α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-[1-(methyl-d3)ethyl-2,2,2-d3]-benzeneacetonitrile
InChI formel InChI=1S/C27H38N2O4/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6/h9-12,17-18,20H,8,13-16H2,1-7H3/i1D3,2D3
SMIL [2H]C([2H])([2H])C(C([2H])([2H])[2H])C(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2
IUPAC navn 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)pentanenitrile
Molekylvægt (g/mol) 460.6386
Formel vægt 460.3208
Procent renhed >95