Få mere at vide
Venlafaxine EP Impurity B (as Hydrochloride), Mikromol™
Artikelnummer. 17980766
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17980766 25 mg
17981586 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17980766 Leverandør Mikromol Leverandørnr. MM0393.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Venlafaxine EP Impurity B (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride
CAS 2174001-92-2
Analyt- eller komponentnavne Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N O3 . Cl H
Certificeringer/compliance ISO 17025
Synonym Venlafaxine Hydrochloride Imp. B (EP), Venlafaxine Imp. B (EP), Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride, Venlafaxine Hydrochloride Imp. B (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity B as Hydrochloride, Venlafaxine Impurity B as Hydrochloride
InChI formel InChI=1S/C14H21NO3.ClH/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11;/h6-9,13H,5,10H2,1-4H3;1H
SMIL Cl.CCOC(=O)C(CN(C)C)c1ccc(OC)cc1
IUPAC navn ethyl 3-(dimethylamino)-2-(4-methoxyphenyl)propanoate;hydrochloride
Molekylvægt (g/mol) 287.78
Mængde 25 mg
Formel vægt 287.1288 g/mol